Nalgene™ PC Low Particulate Biotainers
Thermo Scientific™

Nalgene™ PC Low Particulate Biotainers

Nalgene PC Low Particulate Biotainers are certified clean to USP <788> standards, making them an ideal solution for biological and pharmaceutical manufacturing.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)ShapeCertifications/ComplianceTemperature Range (Metric)
383120-421 LSquareUSP <778>-70°C to 70°C
383030-42125 mLSquareUSP <778>-100°C to 100°C
383233-422 LSquareUSP <778>-100°C to 100°C
383405-425 LSquareUSP Class VI, Non-cytotoxic, USP <661>, Non-pyrogenic, FDA 21 CFR 177.1315-100°C to 100°C
383405-165 LSquare with HandleUSP <778>, USP Class VI, Non-cytotoxic, USP <661>, Non-pyrogenic, FDA 21 CFR 177.1315-100°C to 100°C
383410-4210 LSquareUSP Class VI, Non-cytotoxic, USP <661>, Non-pyrogenic, FDA 21 CFR 177.1520-100°C to 100°C
383410-0810 LSquare with HandleUSP Class VI, Non-cytotoxic, USP <661>, Non-pyrogenic, FDA 21 CFR 177.1520-100°C to 100°C
383423-4220 LSquareUSP Class VI, Non-cytotoxic, USP <661>, Non-pyrogenic, FDA 21 CFR 177.1520-100°C to 100°C
Catalog number 383120-42
Price (KRW)
1,669,000
Case of 35
Capacity (Metric):
1 L
Shape:
Square
Certifications/Compliance:
USP <778>
Temperature Range (Metric):
-70°C to 70°C
Price (KRW)
1,669,000
Case of 35

Specifically designed for pharmaceutical and biological manufacturing, Thermo Scientific™ Nalgene™ Polycarbonate Biotainer™ Bottles and Carboys reduce the risk of carry-over contamination. Durable, translucent PC construction ensures the integrity of your sample, even frozen to -100°C. Provided sterile and ready-to-use; including certified clean products, which meet USP <788> standards and endotoxins below 0.5 EU/mL. Certified clean products include triple layer packaging, with two heat-sealed layers to facilitate ease of entry into cleanrooms.

  • Certified clean products include triple layer packaging, with two heat-sealed layers to facilitate ease of entry into cleanrooms
  • Sterile, ready-to-use products with a Sterility Assurance Level of 10-6 inside the bottle/closure
  • Robust, PC can be used from -100°C to 100°C, allowing for both heated mixing and frozen storage or shipment. Must be tested in actual use conditions as results may vary depending upon application.
  • Inspection procedures throughout manufacturing include leak testing, dimensional testing, visual inspection for particulates and release testing to ensure the product meets all specifications
  • All container/closure systems are certified leakproof** to protect valuable contents
  • Products can be supplied with forced extraction studies*, validation binders* and certificates
  • Customer support includes our highly experienced commercial specialist teams, regulatory team, quality team and regional technical support

Additionally our certified clean products include:

  • Clean room produced in ISO Class 7 to limit particulates during molding
  • Two heat-sealed sterile barriers to protect product prior to use
  • Heat-sealed bags include quick-tears on the top and bottom and placed in carton liner twisted and tape/tie sealed to prevent gross contamination on the inner packaging
  • Certified and lot-to-lot tested to meet USP <788> low particulate requirements to limit potential particulate contamination
  • Certified low endotoxin (0.50 EU/mL) according to USP <85> to reduce pyrogens

*Studies are non-GMP and provided for informational purposes only. The identify and quantity of extractables may vary with routine changes made to the manufacturing process. The studies may assist you in determining whether further leachable studies are warranted to meet any applicable regulatory requirements or expectations, and are available under CDA by contacting us at rocregsupport@thermofisher.com *The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100 mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100 mm after they are filled with water, inverted for 15 minutes, and no water escapes.

Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications
AutoclavableNot Autoclavable
Capacity (English)33.8 oz.
Certifications/ComplianceUSP <778>
Closure Size48 mm
For Use With (Application)Media/Buffer Preparation ​, Media/Buffer Storage, Sampling, Storage, Transportation ​, Validation Testing, Drug Substance, Drug Product
Low ParticulateUSP <788> (Low particulate)
No. per Case35/Cs.
Product TypeStorage and Transport Bottle
ShapeSquare
Temperature Range (Metric)-70°C to 70°C
Workflow StepUpstream media/buffer prep and storage. Upstream process solution storage. Downstream validation, downstream intermediate storage and transport. Bulk drug product/substance transportation.
Capacity (Metric)1 L
ColorTranslucent Blue
MaterialPolycarbonate
SterilitySterile
Unit SizeCase of 35