Nalgene™ PETG Low Particulate Biotainer Bottles
Nalgene™ PETG Low Particulate Biotainer Bottles
Nalgene™ PETG Low Particulate Biotainer Bottles
Thermo Scientific™

Nalgene™ PETG Low Particulate Biotainer Bottles

Nalgene PETG Low Particulate Biotainer Bottles are certified clean to USP <788> standards, making them an excellent solution for biological and pharmaceutical manufacturing.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호용량(미터법)포함수량
383230-422 L, 2 LSide Handle, Side Handle4 x 5 Bottles, 4 x 5 Bottles
383025-42125 mL, 125 mLSide Handle, Side Handle20 x 5 Bottles, 20 x 5 Bottles
383005-42500 mL, 500 mLSide Handle, Side Handle14 x 5 Bottles, 14 x 5 Bottles
383110-421 L, 1 LSide Handle, Side Handle7 x 5 Bottles, 7 x 5 Bottles
383230-202 L, 2 LSide Handle, Side Handle20 Bottles, 20 Bottles
383415-165 L, 5 LSide Handle, Side Handle6 Bottles, 6 Bottles
383415-425 L, 5 LNeck Handle, Neck Handle6 Bottles, 6 Bottles
카탈로그 번호 383230-42
제품 가격(KRW)
1,535,000
Each
용량(미터법):
2 L, 2 L
포함:
Side Handle, Side Handle
수량:
4 x 5 Bottles, 4 x 5 Bottles
제품 가격(KRW)
1,535,000
Each
Specifically designed for pharmaceutical and biological manufacturing, Thermo Scientific™ Nalgene™ Certified Clean PETG Biotainer™ Bottles reduce the risk of carry-over contamination.
  • Specifically designed for pharmaceutical and biological manufacturing
  • Certified Clean, validated to USP <788>
  • <0.5 EU/mL Endotoxin
For Research Use Only. Not for use in diagnostic procedures.
사양
오토클레이브 가능Not Autoclavable
용량(영국식 단위)67.628 fl. oz.
인증/적합성USP <778>
마개 색상White, White
마개 라이너LDPE, LDPE
마개 재질Polypropylene, Polypropylene
마개 크기48 mm, 48 mm
직경(미터법)116 mm
직경(미터법) 외부116 mm
직경(미터법) 외부 넥48 mm, 48 mm
용도(애플리케이션)Media/Buffer Preparation ​, Media/Buffer Preparation ​, Media/Buffer Storage, Media/Buffer Storage, Sampling, Sampling, Storage, Storage, Transportation ​, Transportation ​, Validation Testing, Validation Testing, Drug Substance, Drug Substance, Drug Product, Drug Product
높이(미터법)265 mm, 265 mm
포함Side Handle, Side Handle
길이(영국식 단위)24 in., 24 in.
저 입자USP <788> (Low particulate), USP <788> (Low particulate)
케이스당 수20/Cs., 20/Cs.
팩당 수5/Pk., 5/Pk.
수량4 x 5 Bottles, 4 x 5 Bottles
폭(영국식 단위)19.25 in., 19.25 in.
워크플로우 단계Upstream media/buffer prep and storage. Upstream process solution storage. Downstream validation, downstream intermediate storage and transport. Bulk drug product/substance transportation.
오토클레이브 가능Not Autoclavable
용량(영국식 단위)67.628 fl. oz.
용량(미터법)2 L, 2 L
인증/규정준수USP <778>
색상Clear, Clear
물질PET, PETG
제품 유형Square Bottle, Biotainer
멸균Sterile, Sterile
Unit SizeEach