Bis(2-diphenylphosphinophenyl)ether, 99%
Bis(2-diphenylphosphinophenyl)ether, 99%
Bis(2-diphenylphosphinophenyl)ether, 99%
Thermo Scientific Chemicals

Bis(2-diphenylphosphinophenyl)ether, 99%

CAS: 166330-10-5 | C36H28OP2 | 538.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 383370050
Price (USD)
75.65
Special offer
Online exclusive
Ends: 28-Dec-2026
88.80
Save 13.15
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
75.65
Special offer
Online exclusive
Ends: 28-Dec-2026
88.80
Save 13.15
Each
Chemical Identifiers
CAS166330-10-5
IUPAC Name{2-[2-(diphenylphosphanyl)phenoxy]phenyl}diphenylphosphane
Molecular FormulaC36H28OP2
InChI KeyRYXZOQOZERSHHQ-UHFFFAOYSA-N
SMILESO(C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder or crystals
Melting point184°C to 187°C
Appearance (Color)White to off-white
HPLC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only