Ácido 5-hexinoico, 97 %, Thermo Scientific Chemicals
Ácido 5-hexinoico, 97 %, Thermo Scientific Chemicals
Ácido 5-hexinoico, 97 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ácido 5-hexinoico, 97 %, Thermo Scientific Chemicals

CAS: 53293-00-8 | C6H8O2 | 112.13 g/mol
Have Questions?
Número de catálogoCantidadEnvase
38370025025 gFrasco de vidrio
Número de catálogo 383700250
Precio (USD)
-
Cantidad:
25 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Identificadores químicos
CAS53293-00-8
IUPAC Namehex-5-ynoate
Molecular FormulaC6H7O2
InChI KeyVPFMEXRVUOPYRG-UHFFFAOYSA-M
SMILES[O-]C(=O)CCCC#C
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)Clear yellow
Titration with NaOH>=96.0 %
Infrared spectrumConforms
Refractive index1.4480 to 1.4500 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only