Trimethyl orthopropionate, 97%
Trimethyl orthopropionate, 97%
Trimethyl orthopropionate, 97%
Thermo Scientific Chemicals

Trimethyl orthopropionate, 97%

CAS: 24823-81-2 | C6H14O3 | 134.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 384620250
Price (EUR)
49,10
Each
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
49,10
Each
Chemical Identifiers
CAS24823-81-2
IUPAC Name1,1,1-trimethoxypropane
Molecular FormulaC6H14O3
InChI KeyZGMNAIODRDOMEK-UHFFFAOYSA-N
SMILESCCC(OC)(OC)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless
Infrared spectrumConforms
Refractive index1.3970 to 1.3990 (20°C, 589 nm)
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only