Ammonium thioglycolate, 70 wt.% solution in water
Ammonium thioglycolate, 70 wt.% solution in water
Ammonium thioglycolate, 70 wt.% solution in water
Thermo Scientific Chemicals

Ammonium thioglycolate, 70 wt.% solution in water

Ammonium thioglycolate, 68.0 to 72.0%, C2H7NO2S, CAS Number-5421-46-5, 7732-18-5 | CAS: 5421-46-5 | C2H7NO2S | 109.143 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Catalog number 384682500
Price (USD)
35.65
Online exclusive
39.30
Save 3.65
Each
Quantity:
250 mL
Request bulk or custom format
Price (USD)
35.65
Online exclusive
39.30
Save 3.65
Each
Chemical Identifiers
CAS5421-46-5
IUPAC Nameammonium 2-sulfanylacetate
Molecular FormulaC2H7NO2S
InChI KeyZZTCCAPMZLDHFM-UHFFFAOYSA-N
SMILES[NH4+].[O-]C(=O)CS
View more
SpecificationsSpecification SheetSpecification Sheet
pH5.8 to 6.4
Identificationammonium:positive
Appearance (Form)Solution
Titration Iodimetric68.0 to 72.0 %
Iron (Fe)=<0.2 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only