1-Isopropylpiperazine, 98+%
1-Isopropylpiperazine, 98+%
1-Isopropylpiperazine, 98+%
Thermo Scientific Chemicals

1-Isopropylpiperazine, 98+%

CAS: 4318-42-7 | C7H16N2 | 128.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 384810010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS4318-42-7
IUPAC Name1-(propan-2-yl)piperazine
Molecular FormulaC7H16N2
InChI KeyWHKWMTXTYKVFLK-UHFFFAOYSA-N
SMILESCC(C)N1CCNCC1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.0 %
Refractive index1.4700 to 1.4720 (20°C, 589 nm)
Appearance (Color)Clear light yellow
Infrared spectrumConforms
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only