1-(2-Hydroxyethyl)-4-methylpiperazine, 98%
1-(2-Hydroxyethyl)-4-methylpiperazine, 98%
1-(2-Hydroxyethyl)-4-methylpiperazine, 98%
Thermo Scientific Chemicals

1-(2-Hydroxyethyl)-4-methylpiperazine, 98%

CAS: 5464-12-0 | C7H16N2O | 144.22 g/mol
数量:
1 g
5 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 384830010
価格(JPY)
-
数量:
1 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS5464-12-0
IUPAC Name2-(4-methylpiperazin-1-yl)ethan-1-ol
Molecular FormulaC7H16N2O
InChI KeyQHTUMQYGZQYEOZ-UHFFFAOYSA-N
SMILESCN1CCN(CCO)CC1
さらに表示
Infrared spectrumConforms
Appearance (Form)Low melting solid
Refractive index1.4850 to 1.4870 (20°C, 589 nm)
Appearance (Color)Light yellow
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only