(Trimethylsilyl)diazomethane, 1.8 to 2.4M solution in hexanes
(Trimethylsilyl)diazomethane, 1.8 to 2.4M solution in hexanes
(Trimethylsilyl)diazomethane, 1.8 to 2.4M solution in hexanes
Thermo Scientific Chemicals

(Trimethylsilyl)diazomethane, 1.8 to 2.4M solution in hexanes

CAS: 18107-18-1 | C4H10N2Si | 114.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 385330050
Price (USD)
89.90
Each
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
89.90
Each
Chemical Identifiers
CAS18107-18-1
IUPAC Name(diazomethyl)trimethylsilane
Molecular FormulaC4H10N2Si
InChI KeyONDSBJMLAHVLMI-UHFFFAOYSA-N
SMILESC[Si](C)(C)C=[N+]=[N-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow
Strength1.8 to 2.4 M
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent
RUO – Research Use Only