2-Mercaptophenol, 98+%
2-Mercaptophenol, 98+%
2-Mercaptophenol, 98+%
2-Mercaptophenol, 98+%
Thermo Scientific Chemicals

2-Mercaptophenol, 98+%

CAS: 1121-24-0 | C6H6OS | 126.18 g/mol
数量:
1 g
5 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 385600050
価格(JPY)
-
見積もりを依頼する
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1121-24-0
IUPAC Name2-sulfanylphenol
Molecular FormulaC6H6OS
InChI KeyVMKYTRPNOVFCGZ-UHFFFAOYSA-N
SMILESOC1=CC=CC=C1S
さらに表示
Appearance (Color)Clear yellow to light green
Appearance (Form)Liquid
GC>=98.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only