Glycine, 99%, ACS reagent
Glycine, 99%, ACS reagent
Glycine, 99%, ACS reagent
Thermo Scientific Chemicals

Glycine, 99%, ACS reagent

CAS: 56-40-6 | C2H5NO2 | 75.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
Plastic Bottle
Catalog number 385900500
Price (TWD)
-
Quantity:
50 g
Packaging:
Plastic Bottle
Request bulk or custom format
Chemical Identifiers
CAS56-40-6
IUPAC Name2-aminoacetic acid
Molecular FormulaC2H5NO2
InChI KeyDHMQDGOQFOQNFH-UHFFFAOYSA-N
SMILESNCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Sulfate (SO4)=<0.005 %
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with HClO4>=98.5 %
Sulfated ash=<0.1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only