Isopropylmagnesium chloride - Lithium chloride complex, 1.3M solution in THF, AcroSeal™
Isopropylmagnesium chloride - Lithium chloride complex, 1.3M solution in THF, AcroSeal™
Isopropylmagnesium chloride - Lithium chloride complex, 1.3M solution in THF, AcroSeal™
Thermo Scientific Chemicals

Isopropylmagnesium chloride - Lithium chloride complex, 1.3M solution in THF, AcroSeal™

CAS: 745038-86-2 | C3H7Cl2LiMg | 145.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
View all
AcroSeal™ Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 386281000
Price (EUR)
113,65
Online Exclusive
126,00
Save 12,35 (10%)
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (EUR)
113,65
Online Exclusive
126,00
Save 12,35 (10%)
Each
Chemical Identifiers
CAS745038-86-2
IUPAC Namelithium(1+) chloro(propan-2-yl)magnesium chloride
Molecular FormulaC3H7Cl2LiMg
InChI KeyCWTUREABAILGIK-UHFFFAOYSA-L
SMILES[Li+].[Cl-].CC(C)[Mg]Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Appearance (Color)Clear yellow to grey to brown
AppearanceMay develop some turbidity or precipitate
Titration Grignard1.1 to 1.5 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only