Tetrafluoroborato de triciclohexilfosfonio, 99 %, Thermo Scientific Chemicals
Tetrafluoroborato de triciclohexilfosfonio, 99 %, Thermo Scientific Chemicals
Tetrafluoroborato de triciclohexilfosfonio, 99 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetrafluoroborato de triciclohexilfosfonio, 99 %, Thermo Scientific Chemicals

CAS: 58656-04-5 | C18H34BF4P | 368.25 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
1 g
5 g
Envase:
Frasco de vidrio
Número de catálogo 386830050
Precio (EUR)
101,00
Each
Cantidad:
5 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Precio (EUR)
101,00
Each
Identificadores químicos
CAS58656-04-5
IUPAC Nametetrafluoroboranuide; tricyclohexylphosphanium
Molecular FormulaC18H34BF4P
InChI KeyMYSMMEUXKHJYKH-UHFFFAOYSA-O
SMILESF[B-](F)(F)F.C1CCC(CC1)[PH+](C1CCCCC1)C1CCCCC1
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Infrared spectrumConforms
Elemental Analysis>=98.5 %
Appearance (Form)Powder
Appearance (Color)White
Melting point165°C to 169°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only