1-Bromo-2-butyne, 98%
1-Bromo-2-butyne, 98%
1-Bromo-2-butyne, 98%
Thermo Scientific Chemicals

1-Bromo-2-butyne, 98%

CAS: 3355-28-0 | C4H5Br | 132.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
View all
Glass Bottle
Catalog number 387020250
Price (EUR)
498,00
Each
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (EUR)
498,00
Each
Chemical Identifiers
CAS3355-28-0
IUPAC Name1-bromobut-2-yne
Molecular FormulaC4H5Br
InChI KeyLNNXOEHOXSYWLD-UHFFFAOYSA-N
SMILESCC#CCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear yellow-green
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.5070 to 1.5090 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only