Sodium sulfide nonahydrate, 98+%, extra pure, Thermo Scientific Chemicals
Sodium sulfide nonahydrate, 98+%, extra pure, Thermo Scientific Chemicals
Sodium sulfide nonahydrate, 98+%, extra pure, Thermo Scientific Chemicals
Sodium sulfide nonahydrate, 98+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium sulfide nonahydrate, 98+%, extra pure, Thermo Scientific Chemicals

数量:
500 g
2.5 kg
パッケージ:
Glass bottle
製品番号(カタログ番号) 387065000
価格(JPY)
-
数量:
500 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1313-84-4
IUPAC Namedisodium nonahydrate sulfanediide
Molecular FormulaH18Na2O9S
InChI KeyZGHLCBJZQLNUAZ-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--]
さらに表示
Appearance (Color)Colorless to white to light yellow
Appearance (Form)Powder or crystals and/or chunks
Titration Iodimetric32.0 to 38.0 % (Na2S)
Ammonium (NH4)=<0.005 %
Sulfite (as SO2)=<2000 ppm
さらに表示

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only