2-Hydroxyphenylboronic acid, 97%
2-Hydroxyphenylboronic acid, 97%
2-Hydroxyphenylboronic acid, 97%
Thermo Scientific Chemicals

2-Hydroxyphenylboronic acid, 97%

CAS: 89466-08-0 | C6H7BO3 | 137.93 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
1 g
5 g
產品號碼 387500010
價格 (TWD)
-
Quantity:
1 g
Request bulk or custom format
化學識別
CAS89466-08-0
IUPAC Name(2-hydroxyphenyl)boronic acid
Molecular FormulaC6H7BO3
InChI KeyYDMRDHQUQIVWBE-UHFFFAOYSA-N
SMILESOB(O)C1=CC=CC=C1O
檢視更多
規格Specification Sheet規格表
Appearance (Color)White
Infrared spectrumConforms
HPLC>=96.0 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only