Poly(ethylene glycol methyl ether), average M.W. 500
Poly(ethylene glycol methyl ether), average M.W. 500
Poly(ethylene glycol methyl ether), average M.W. 500
Thermo Scientific Chemicals

Poly(ethylene glycol methyl ether), average M.W. 500

CAS: 9004-74-4 | (C2H4O)nCH4O | 76.10 g/mol
数量:
500 g
1 kg
パッケージ:
Glass bottle
製品番号(カタログ番号) 387890010
価格(JPY)
-
数量:
1 kg
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS9004-74-4
Molecular Formula(C2H4O)nCH4O
InChI KeyXNWFRZJHXBZDAG-UHFFFAOYSA-N
SMILESCOCCO
MDL NumberMFCD00084416
Water=<1.0 %
pH5.0 to 7.5 (1 % in water)
Appearance (Color)Colorless or white
Infrared spectrumConforms
Average molecular weight450 to 550
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only