Tetrahydro-2H-pyran-2-ol, 90%, cyclized form of 5-Hydroxypentanal
Tetrahydro-2H-pyran-2-ol, 90%, cyclized form of 5-Hydroxypentanal
Tetrahydro-2H-pyran-2-ol, 90%, cyclized form of 5-Hydroxypentanal
Thermo Scientific Chemicals

Tetrahydro-2H-pyran-2-ol, 90%, cyclized form of 5-Hydroxypentanal

CAS: 694-54-2 | C5H10O2 | 102.13 g/mol
Have Questions?
Catalog NumberQuantityPackaging
38790025025 gGlass bottle
Catalog number 387900250
Price (USD)
264.65
Special offer
Online exclusive
Ends: 28-Dec-2026
311.00
Save 46.35
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
264.65
Special offer
Online exclusive
Ends: 28-Dec-2026
311.00
Save 46.35
Each
Chemical Identifiers
CAS694-54-2
IUPAC Nameoxan-2-ol
Molecular FormulaC5H10O2
InChI KeyCELWCAITJAEQNL-UHFFFAOYNA-N
SMILESOC1CCCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=88 %
Appearance (Color)Clear colorless
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only