Soda lime, pure, granular, with indicator white to violet
Soda lime, pure, granular, with indicator white to violet
Soda lime, pure, granular, with indicator white to violet
Soda lime, pure, granular, with indicator white to violet
Acros Organics

Soda lime, pure, granular, with indicator white to violet

CAS: 8006-28-8 | CaHNaO2 | 96.074 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Packaging:
Glass Bottle
Catalog number 388350500
HSN 28259090
Price (INR)
3,835.00
Each
Add to cart
Quantity:
50 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (INR)
3,835.00
Each
Add to cart
Chemical Identifiers
CAS8006-28-8
IUPAC Namesodium oxocalcium hydroxide
Molecular FormulaCaHNaO2
InChI KeyHUAUNKAZQWMVFY-UHFFFAOYSA-M
SMILES[OH-].[Na+].O=[Ca]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Granules
CO2 absorption capacity>=19.0 %
IndicatorPasses test
Appearance (Color)White to beige to light grey
Loss on drying=<20 % (10 g, 200°C, 2 h)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Gas Adsorbent, Indicator
RUO – Research Use Only