Lithium phenoxide, 0.6M solution in THF, AcroSeal™
Lithium phenoxide, 0.6M solution in THF, AcroSeal™
Lithium phenoxide, 0.6M solution in THF, AcroSeal™
Lithium phenoxide, 0.6M solution in THF, AcroSeal™
Thermo Scientific Chemicals

Lithium phenoxide, 0.6M solution in THF, AcroSeal™

CAS: 555-24-8 | C6H5LiO | 100.05 g/mol
Have Questions?
Catalog NumberQuantityPackaging
388611000100 mLAcroSeal™ Glass Bottle
Catalog number 388611000
Price (USD)
123.00
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)
123.00
Each
Chemical Identifiers
CAS555-24-8
IUPAC Namelithium(1+) benzenolate
Molecular FormulaC6H5LiO
InChI KeyXAVQZBGEXVFCJI-UHFFFAOYSA-M
SMILES[Li+].[O-]C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow to orange
Titration with HClO40.55 to 0.65 M
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only