Hydrogen chloride, 1M solution in ethyl acetate
Hydrogen chloride, 1M solution in ethyl acetate
Hydrogen chloride, 1M solution in ethyl acetate
Thermo Scientific Chemicals

Hydrogen chloride, 1M solution in ethyl acetate

CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Catalog NumberQuantity
388701000100 mL
Catalog number 388701000
Price (USD)
67.70
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
67.70
Each
Chemical Identifiers
CAS7647-01-0
IUPAC Namehydrogen chloride
Molecular FormulaClH
InChI KeyVEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILESCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Strength0.95 to 1.15 M
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only