1,1-Dimethylurea, 98+%
1,1-Dimethylurea, 98+%
1,1-Dimethylurea, 98+%
Thermo Scientific Chemicals

1,1-Dimethylurea, 98+%

CAS: 598-94-7 | C3H8N2O | 88.11 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
產品號碼 388740050
價格 (TWD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS598-94-7
IUPAC Name1,1-dimethylurea
Molecular FormulaC3H8N2O
InChI KeyYBBLOADPFWKNGS-UHFFFAOYSA-N
SMILESCN(C)C(N)=O
檢視更多
規格Specification Sheet規格表
Infrared spectrumConforms
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
Melting point179°C to 183°C
Titration with HClO4>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only