Benzyl 2-bromoethyl ether, 97%
Benzyl 2-bromoethyl ether, 97%
Benzyl 2-bromoethyl ether, 97%
Thermo Scientific Chemicals

Benzyl 2-bromoethyl ether, 97%

CAS: 1462-37-9 | C9H11BrO | 215.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 388871000
Price (TWD)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS1462-37-9
IUPAC Name[(2-bromoethoxy)methyl]benzene
Molecular FormulaC9H11BrO
InChI KeyFWOHDAGPWDEWIB-UHFFFAOYSA-N
SMILESBrCCOCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=96.0 %
Appearance (Form)Liquid
Refractive index1.5390 to 1.5420 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only