Ethynylmagnesium chloride, 0.5M solution in THF/Toluene, AcroSeal™
Ethynylmagnesium chloride, 0.5M solution in THF/Toluene, AcroSeal™
Ethynylmagnesium chloride, 0.5M solution in THF/Toluene, AcroSeal™
Thermo Scientific Chemicals

Ethynylmagnesium chloride, 0.5M solution in THF/Toluene, AcroSeal™

CAS: 65032-27-1 | C2HClMg | 84.79 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
View all
AcroSeal™ Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 388951000
Price (EUR)
58,65
Online exclusive
64,80
Save 6,15 (9%)
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (EUR)
58,65
Online exclusive
64,80
Save 6,15 (9%)
Each
Chemical Identifiers
CAS65032-27-1
IUPAC Namechloro(ethynyl)magnesium
Molecular FormulaC2HClMg
InChI KeyYEWNLGTVYUADOR-UHFFFAOYSA-M
SMILESCl[Mg]C#C
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Grignard4 to 6 % (As ethynyl-MgCl)
Appearance (Color)Yellow to dark brown
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only