Manganese(II) acetate, 98+%, anhydrous
Manganese(II) acetate, 98+%, anhydrous
Manganese(II) acetate, 98+%, anhydrous
Thermo Scientific Chemicals

Manganese(II) acetate, 98+%, anhydrous

CAS: 638-38-0 | C4H6MnO4 | 173.03 g/mol
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 389140050
Price (JPY)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS638-38-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Beige to pink to grey to light brown
Appearance (Form)Powder
Infrared spectrumConforms
Titration Complexometric98.0 to 102.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only