N,O-Bis(trimethylsilyl)trifluoroacetamide, with 1% trimethylsilyl chloride
N,O-Bis(trimethylsilyl)trifluoroacetamide, with 1% trimethylsilyl chloride
N,O-Bis(trimethylsilyl)trifluoroacetamide, with 1% trimethylsilyl chloride
Acros Organics

N,O-Bis(trimethylsilyl)trifluoroacetamide, with 1% trimethylsilyl chloride

CAS: 25561-30-2 | C8H18F3NOSi2 | 257.4 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Catalog number 389321000
HSN 29319090
Price (INR)
64,603.00
Each
Quantity:
100 g
Request bulk or custom format
Price (INR)
64,603.00
Each
Chemical Identifiers
CAS25561-30-2
IUPAC Nametrimethylsilyl 2,2,2-trifluoro-N-(trimethylsilyl)ethanimidate
Molecular FormulaC8H18F3NOSi2
InChI KeyXCOBLONWWXQEBS-UHFFFAOYSA-N
SMILESC[Si](C)(C)OC(=N[Si](C)(C)C)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
GC=<1.5 % TMCS
GC>=98.0 % (excluding TMCS)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Silylation reagent
RUO – Research Use Only