Zinc sulfate monohydrate, 99%
Zinc sulfate monohydrate, 99%
Zinc sulfate monohydrate, 99%
Thermo Scientific Chemicals

Zinc sulfate monohydrate, 99%

CAS: 7446-19-7 | O4SZn·H2O | 179.47 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Catalog number 389802500
Price (USD)
47.10
Each
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
47.10
Each
Chemical Identifiers
CAS7446-19-7
IUPAC Namezinc(2+) hydrate sulfate
Molecular FormulaH2O5SZn
InChI KeyRNZCSKGULNFAMC-UHFFFAOYSA-L
SMILESO.[Zn++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder
Titration Complexometric98.5 to 101.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only