N-BOC-1,2-diaminoethane, 98%
N-BOC-1,2-diaminoethane, 98%
N-BOC-1,2-diaminoethane, 98%
Thermo Scientific Chemicals

N-BOC-1,2-diaminoethane, 98%

CAS: 57260-73-8 | C7H16N2O2 | 160.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass Bottle
Catalog number 389900010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS57260-73-8
IUPAC Nametert-butyl N-(2-aminoethyl)carbamate
Molecular FormulaC7H16N2O2
InChI KeyAOCSUUGBCMTKJH-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)NCCN
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Form)Viscous liquid
Refractive index1.4550 to 1.4570 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only