(S)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazine, 97+%
(S)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazine, 97+%
(S)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazine, 97+%
Thermo Scientific Chemicals

(S)-2,5-Dihydro-3,6-dimethoxy-2-isopropylpyrazine, 97+%

(S)-2, 5-Dihydro-3, 6-dimethoxy-2-isopropylpyrazine, 97+%, C9H16N2O2, CAS Number-78342-42-4 | CAS: 78342-42-4 | C9H16N2O2 | 184.24 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3899600505 mLGlass bottle
Catalog number 389960050
Price (USD)
-
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS78342-42-4
IUPAC Name3,6-dimethoxy-2-(propan-2-yl)-2,5-dihydropyrazine
Molecular FormulaC9H16N2O2
InChI KeyFCFWEOGTZZPCTO-UHFFFAOYNA-N
SMILESCOC1=NC(C(C)C)C(OC)=NC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Specific optical rotation+96° to +109° (20°C, 589 nm) (C=1, EtOH)
Appearance (Form)Liquid
Refractive index1.4600 to 1.4620 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only