Invitrogen™

IgG1 Mouse ELISA Standard

Catalog number: 39-50410-65
Invitrogen™

IgG1 Mouse ELISA Standard

Catalog number: 39-50410-65
Catalog Number
39-50410-65
Unit Size
4 pack
Price (USD)
Price: 207.00
Your price:
Quantity
Catalog NumberUnit SizePrice (USD)Quantity
39-50410-654 pack
Price: 207.00
Your price:
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
This product is a component of a respective eBioscience Kit. Please refer to the relevant manual for each product on our website.

IgG is the major immunoglobulin in blood, lymph fluid, cerebrospinal fluid, and peritoneal fluid and a key player in the humoral immune response.

The Fc portion of IgG, but not F(ab')2 or Fab fragments, can cross the placenta of a mother to enter the fetal circulation providing the fetus with postpartum protection.

IgG molecules are able to react with Fc-gamma receptors that are present on the surfaces of macrophages, neutrophils, natural killer cells, and can activate the complement system.

The binding of the Fc portion of IgG to the receptor present on a phagocyte is a critical step in the opsonizing property IgG provides to the immune response. Phagocytosis of particles coated with IgG antibodies is a vital mechanism to cope with microorganisms.

IgG is produced in a delayed response to an infection and can be retained in the body for a long time. The longevity in serum makes IgG most useful for passive immunization by transfer of this antibody. Detection of IgG usually indicates a prior infection or vaccination.

Reported Application
ELISA Recombinant Standard, ELISA

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format
Kit
Quantity
4 Pack
Recombinant
Recombinant
For Use With (Equipment)
Microplate Reader
Former Identifiers
eBioscience
Immunoassay Kit Format
ELISA Standard
Target Species Primary
Mouse
Target Kit Named
IgG1
Target Species Named
Mouse
Target Species Validated
Mouse
Form
Liquid

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text