4-Bromo-1-butanol, 85+%
4-Bromo-1-butanol, 85+%
4-Bromo-1-butanol, 85+%
Thermo Scientific Chemicals

4-Bromo-1-butanol, 85+%

CAS: 33036-62-3 | C4H9BrO | 153.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 390010010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS33036-62-3
IUPAC Name4-bromobutan-1-ol
Molecular FormulaC4H9BrO
InChI KeySIJLYRDVTMMSIP-UHFFFAOYSA-N
SMILESOCCCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to purple to brown
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=85.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only