Isopropyl ether, for analysis, stabilized
Isopropyl ether, for analysis, stabilized
Isopropyl ether, for analysis, stabilized
Thermo Scientific Chemicals

Isopropyl ether, for analysis, stabilized

CAS: 108-20-3 | C6H14O | 102.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 390660010
Price (USD)
83.65
Special offer
Online exclusive
Ends: 28-Dec-2026
97.80
Save 14.15
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
83.65
Special offer
Online exclusive
Ends: 28-Dec-2026
97.80
Save 14.15
Each
Chemical Identifiers
CAS108-20-3
IUPAC Name2-(propan-2-yloxy)propane
Molecular FormulaC6H14O
InChI KeyZAFNJMIOTHYJRJ-UHFFFAOYSA-N
SMILESCC(C)OC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Magnesium (Mg)=<0.1 ppm
Lead (Pb)=<0.1 ppm
Zinc (Zn)=<0.1 ppm
Appearance (Form)Clear liquid
Water=<0.1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only