N,N-Dimethylacetamide, 99.5+%, for analysis
N,N-Dimethylacetamide, 99.5+%, for analysis
N,N-Dimethylacetamide, 99.5+%, for analysis
Thermo Scientific Chemicals

N,N-Dimethylacetamide, 99.5+%, for analysis

CAS: 127-19-5 | C4H9NO | 87.12 g/mol
Catalog number 390800010
Price (JPY)
-
Request bulk or custom format
Chemical Identifiers
CAS127-19-5
IUPAC NameN,N-dimethylacetamide
Molecular FormulaC4H9NO
InChI KeyFXHOOIRPVKKKFG-UHFFFAOYSA-N
SMILESCN(C)C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Chloride (Cl)=<0.001 %
Sulfate (SO4)=<0.001 %
Cadmium (Cd)=<0.05 ppm
Calcium (Ca)=<0.5 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only