Thermo Scientific Chemicals

Cesium acetate, 99%, for analysis, Thermo Scientific Chemicals

Catalog number: 390830250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Cesium acetate, 99%, for analysis, Thermo Scientific Chemicals

Catalog number: 390830250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 390830250
Price (EUR)
Quantity
-

Chemical Identifiers

CAS
3396-11-0
IUPAC Name
caesium(1+) acetate
Molecular Formula
C2H3CsO2
InChI Key
ZOAIGCHJWKDIPJ-UHFFFAOYSA-M
SMILES
[Cs+].CC([O-])=O
Potassium (K)
=<20 ppm
Rubidium (Rb)
=<0.12 %
Magnesium (Mg)
=<3 ppm
Calcium (Ca)
=<40 ppm
Aluminium (Al)
=<6 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text

    Safety and Handling


    Classification of the substance or mixture
    CLP classification - Regulation(EC) No 1272/2008
    Reproductive Toxicity
    Category 2
    Label Elements
    Signal Word

    Warning

    Hazard Statements

    H361 - Suspected of damaging fertility or the unborn child

    Precautionary Statements

    P201 - Obtain special instructions before use

    P308 + P311 - IF exposed or concerned: Call a POISON CENTER or doctor