Benzyl alcohol, for analysis
Benzyl alcohol, for analysis
Benzyl alcohol, for analysis
Thermo Scientific Chemicals

Benzyl alcohol, for analysis

CAS: 100-51-6 | C7H8O | 108.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 391220025
Price (USD)
271.00
Each
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
271.00
Each
Chemical Identifiers
CAS100-51-6
IUPAC Namephenylmethanol
Molecular FormulaC7H8O
InChI KeyWVDDGKGOMKODPV-UHFFFAOYSA-N
SMILESOCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
GC>=99.5 %
Aluminium (Al)=<0.5 ppm
Cadmium (Cd)=<0.05 ppm
Chromium (Cr)=<0.02 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only