4-Amino-3-methoxybenzoic acid, 98%
4-Amino-3-methoxybenzoic acid, 98%
4-Amino-3-methoxybenzoic acid, 98%
Thermo Scientific Chemicals

4-Amino-3-methoxybenzoic acid, 98%

CAS: 2486-69-3 | C8H9NO3 | 167.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 391460050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2486-69-3
IUPAC Name4-amino-3-methoxybenzoic acid
Molecular FormulaC8H9NO3
InChI KeyJNFGLYJROFAOQP-UHFFFAOYSA-N
SMILESCOC1=CC(=CC=C1N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Melting point186°C to 191°C
Appearance (Color)White to light beige to grey
Infrared spectrumConforms
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only