4-Hydroxybenzoic acid, sodium salt, 98%
4-Hydroxybenzoic acid, sodium salt, 98%
4-Hydroxybenzoic acid, sodium salt, 98%
Thermo Scientific Chemicals

4-Hydroxybenzoic acid, sodium salt, 98%

CAS: 114-63-6 | C7H5NaO3 | 160.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 391521000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS114-63-6
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=97.5 %
Appearance (Color)White to light beige
Infrared spectrumConforms
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only