Ammonium thiosulfate, 96%, pure
Ammonium thiosulfate, 96%, pure
Ammonium thiosulfate, 96%, pure
Thermo Scientific Chemicals

Ammonium thiosulfate, 96%, pure

CAS: 7783-18-8 | H8N2O3S2 | 148.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
10 kg
Packaging:
View all
Plastic bottle
Catalog number 391660025
Price (USD)
-
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS7783-18-8
IUPAC Namedihydroxy-1λ⁶-disulfen-1-one diamine
Molecular FormulaH8N2O3S2
InChI KeyXYXNTHIYBIDHGM-UHFFFAOYSA-N
SMILESN.N.OS(O)(=O)=S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals and/or chunks
Titration Iodimetric>=95 %
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only