4-Chlorobenzyl bromide, 98%
4-Chlorobenzyl bromide, 98%
4-Chlorobenzyl bromide, 98%
Thermo Scientific Chemicals

4-Chlorobenzyl bromide, 98%

CAS: 622-95-7 | C7H6BrCl | 205.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 391710250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS622-95-7
IUPAC Name1-(bromomethyl)-4-chlorobenzene
Molecular FormulaC7H6BrCl
InChI KeyKQNBRMUBPRGXSL-UHFFFAOYSA-N
SMILESClC1=CC=C(CBr)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point46°C to 51°C
Appearance (Form)Crystals and/or chunks or fused solid
GC>=97.5 %
Appearance (Color)White to almost white
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only