2-Iodobenzonitrile, 99%
2-Iodobenzonitrile, 99%
2-Iodobenzonitrile, 99%
2-Iodobenzonitrile, 99%
Thermo Scientific Chemicals

2-Iodobenzonitrile, 99%

CAS: 4387-36-4 | C7H4IN | 229.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 391800050
Price (EUR)
86,50
Each
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
86,50
Each
Add to cart
Chemical Identifiers
CAS4387-36-4
IUPAC Name2-iodobenzonitrile
Molecular FormulaC7H4IN
InChI KeyJDDAFHUEOVUDFJ-UHFFFAOYSA-N
SMILESIC1=CC=CC=C1C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals and/or chunks
Melting point51°C to 55°C
Infrared spectrumConforms
GC>=98.5 %
Appearance (Color)White to light brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only