Thermo Scientific Chemicals

Silicagel orange, for drying purposes, non toxic grade, Thermo Scientific Chemicals

Catalog number: 392030050
5 kg, Plastic Drum, Each
Thermo Scientific Chemicals

Silicagel orange, for drying purposes, non toxic grade, Thermo Scientific Chemicals

Catalog number: 392030050
5 kg, Plastic Drum, Each
Quantity
Packaging
Catalog number: 392030050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7631-86-9
IUPAC Name
silanedione
Molecular Formula
O2Si
InChI Key
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILES
O=[Si]=O
Appearance (Form)
Beads
Water adsorption
>=20 % (50% rH, 1 bar, 25°C)
Color
Orange (dry) to Colorless (wet)
Appearance (Color)
Orange
Residual water
=<2 % (150°C)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Drying agent
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text