4-Iodobiphenyl, 97+%
4-Iodobiphenyl, 97+%
4-Iodobiphenyl, 97+%
4-Iodobiphenyl, 97+%
Thermo Scientific Chemicals

4-Iodobiphenyl, 97+%

CAS: 1591-31-7 | C12H9I | 280.11 g/mol
数量:
1 g
5 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 392390050
価格(JPY)
-
見積もりを依頼する
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1591-31-7
IUPAC Name4-iodo-1,1'-biphenyl
Molecular FormulaC12H9I
InChI KeyNXYICUMSYKIABQ-UHFFFAOYSA-N
SMILESIC1=CC=C(C=C1)C1=CC=CC=C1
さらに表示
Appearance (Color)Off-white
Appearance (Form)Crystalline powder
Melting point110°C to 114°C
Infrared spectrumConforms
HPLC>=97.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only