Citric acid, diammonium salt, 98%, pure
Citric acid, diammonium salt, 98%, pure
Citric acid, diammonium salt, 98%, pure
Thermo Scientific Chemicals

Citric acid, diammonium salt, 98%, pure

CAS: 3012-65-5 | C6H14N2O7 | 226.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 392505000
Price (USD)
54.65
Special offer
Online exclusive
Ends: 28-Dec-2026
63.60
Save 8.95
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
54.65
Special offer
Online exclusive
Ends: 28-Dec-2026
63.60
Save 8.95
Each
Chemical Identifiers
CAS3012-65-5
IUPAC Namediammonium 3-carboxy-3-hydroxypentanedioate
Molecular FormulaC6H14N2O7
InChI KeyYXVFQADLFFNVDS-UHFFFAOYSA-N
SMILES[NH4+].[NH4+].OC(=O)C(O)(CC([O-])=O)CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Crystalline powder or crystals and/or chunks
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only