2-Hydroxy-5-methylbenzaldehyde, 99%
2-Hydroxy-5-methylbenzaldehyde, 99%
2-Hydroxy-5-methylbenzaldehyde, 99%
Thermo Scientific Chemicals

2-Hydroxy-5-methylbenzaldehyde, 99%

CAS: 613-84-3 | C8H8O2 | 136.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 392630010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS613-84-3
IUPAC Name2-hydroxy-5-methylbenzaldehyde
Molecular FormulaC8H8O2
InChI KeyILEIUTCVWLYZOM-UHFFFAOYSA-N
SMILESCC1=CC=C(O)C(C=O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point53°C to 57°C
Appearance (Form)Crystalline powder
GC>=98.5 %
Appearance (Color)Yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only