Methyl 6-bromohexanoate, 96+%
Methyl 6-bromohexanoate, 96+%
Methyl 6-bromohexanoate, 96+%
Thermo Scientific Chemicals

Methyl 6-bromohexanoate, 96+%

CAS: 14273-90-6 | C7H13BrO2 | 209.08 g/mol
数量:
5 g
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 392690050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS14273-90-6
IUPAC Namemethyl 6-bromohexanoate
Molecular FormulaC7H13BrO2
InChI KeyKYLVAMSNNZMHSX-UHFFFAOYSA-N
SMILESCOC(=O)CCCCCBr
さらに表示
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=96.0 %
Refractive index1.4630 to 1.4650 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only