Methyl 6-bromohexanoate, 96+%
Methyl 6-bromohexanoate, 96+%
Methyl 6-bromohexanoate, 96+%
Thermo Scientific Chemicals

Methyl 6-bromohexanoate, 96+%

CAS: 14273-90-6 | C7H13BrO2 | 209.08 g/mol
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 392691000
Price (JPY)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS14273-90-6
IUPAC Namemethyl 6-bromohexanoate
Molecular FormulaC7H13BrO2
InChI KeyKYLVAMSNNZMHSX-UHFFFAOYSA-N
SMILESCOC(=O)CCCCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=96.0 %
Refractive index1.4630 to 1.4650 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only