2-Amino-5-methylthiazole, 98%
2-Amino-5-methylthiazole, 98%
2-Amino-5-methylthiazole, 98%
Thermo Scientific Chemicals

2-Amino-5-methylthiazole, 98%

CAS: 7305-71-7 | C4H6N2S | 114.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 392790050
Price (USD)
23.65
Special offer
Online exclusive
Ends: 28-Dec-2026
27.40
Save 3.75
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
23.65
Special offer
Online exclusive
Ends: 28-Dec-2026
27.40
Save 3.75
Each
Chemical Identifiers
CAS7305-71-7
IUPAC Name5-methyl-1,3-thiazol-2-amine
Molecular FormulaC4H6N2S
InChI KeyGUABFMPMKJGSBQ-UHFFFAOYSA-N
SMILESCC1=CN=C(N)S1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Powder
GC>=97.5 %
Appearance (Color)Brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only