Thermo Scientific Chemicals

Tetramethylammonium triacetoxyborohydride, 90+%, Thermo Scientific Chemicals

Catalog number: 392850500
50 g, Each
Thermo Scientific Chemicals

Tetramethylammonium triacetoxyborohydride, 90+%, Thermo Scientific Chemicals

Catalog number: 392850500
50 g, Each
Quantity

Chemical Identifiers

CAS
109704-53-2
IUPAC Name
bis(acetyloxy)boranuidyl acetate; tetramethylazanium
Molecular Formula
C10H22BNO6
InChI Key
LCFZZOGKVOTFPU-UHFFFAOYSA-N
SMILES
C[N+](C)(C)C.CC(=O)O[BH-](OC(C)=O)OC(C)=O
Infrared spectrum
Conforms
Appearance (Color)
White
Appearance (Form)
Powder
Titration with HClO4
>=90.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text