Ammonium fluoride, 98+%, for analysis
Ammonium fluoride, 98+%, for analysis
Ammonium fluoride, 98+%, for analysis
Thermo Scientific Chemicals

Ammonium fluoride, 98+%, for analysis

CAS: 12125-01-8 | FH4N | 37.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 393190500
Price (USD)
50.65
Online Exclusive
56.50
Save 5.85 (10%)
Each
Quantity:
50 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
50.65
Online Exclusive
56.50
Save 5.85 (10%)
Each
Chemical Identifiers
CAS12125-01-8
IUPAC Nameamine hydrofluoride
Molecular FormulaFH4N
InChI KeyLDDQLRUQCUTJBB-UHFFFAOYSA-N
SMILESN.F
View more
SpecificationsSpecification SheetSpecification Sheet
Titration excess NaOH>=98.0 %
(NH4)HF2=<1.0 %
Appearance (Form)Adhering crystalline powder
Insoluble matter=<0.005 % (in water)
Sulfate (SO4)=<0.005 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only