Butyraldehyde diethylacetal, 97%
Butyraldehyde diethylacetal, 97%
Butyraldehyde diethylacetal, 97%
Thermo Scientific Chemicals

Butyraldehyde diethylacetal, 97%

CAS: 3658-95-5 | C8H18O2 | 146.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 393430050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3658-95-5
IUPAC Name1,1-diethoxybutane
Molecular FormulaC8H18O2
InChI KeyUVHXZFGCCJLFMX-UHFFFAOYSA-N
SMILESCCCC(OCC)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=96.0 %
Refractive index1.3950 to 1.3980 (20°C, 589 nm)
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only